Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282460.1 | NP_001269389.1 | 5 | 1228 | |
XM_011534989.2 | XP_011533291.1 | 5 | 1378 | ||
XM_011534990.2 | XP_011533292.1 | 5 | 1378 | ||
XM_011534992.2 | XP_011533294.1 | 5 | 1378 | ||
XM_011534993.1 | XP_011533295.1 | 5 | 1378 | ||
XM_017020421.1 | XP_016875910.1 | 5 | 1378 | ||
XM_017020422.1 | XP_016875911.1 | 4 | 1134 | ||
XM_017020423.1 | XP_016875912.1 | 4 | 1548 | ||
XM_017020424.1 | XP_016875913.1 | 5 | 1378 | ||
XM_017020425.1 | XP_016875914.1 | 5 | 1378 | ||
XM_017020426.1 | XP_016875915.1 | 5 | 1378 | ||
XM_017020427.1 | XP_016875916.1 | 5 | 1378 | ||
XM_017020428.1 | XP_016875917.1 | 5 | 1252 | ||
XM_017020429.1 | XP_016875918.1 | 4 | 700 | ||
XM_017020430.1 | XP_016875919.1 | 4 | 574 | ||
XM_017020431.1 | XP_016875920.1 | 3 | 456 | ||
GenBank mRNA | BC030276.1 | - | 5 | 1223 |