Gene Transcripts
- Gene Symbol
- PACS2
- Entrez Gene ID
- 23241
- Gene Name
- phosphofurin acidic cluster sorting protein 2
- Gene Aliases
- PACS-2, PACS1L
- Chromosome Location
- Chr. 14: 105300811 - 105398147 on Build GRCh38
- UniGene ID
- Hs.525626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100913.2 |
NP_001094383.2
|
5, 6 |
1096 |
|
NM_001243127.2 |
NP_001230056.1
|
5, 6 |
642 |
|
NM_015197.3 |
NP_056012.2
|
5, 6 |
1096 |
|
XM_006720090.1 |
XP_006720153.1
|
5, 6 |
761 |
|
XM_006720091.1 |
XP_006720154.1
|
5, 6 |
761 |
|
XM_006720092.2 |
XP_006720155.1
|
5, 6 |
761 |
|
XM_017021105.1 |
XP_016876594.1
|
5, 6 |
761 |
|
XM_017021106.1 |
XP_016876595.1
|
5, 6 |
761 |
|
XM_017021107.1 |
XP_016876596.1
|
5, 6 |
761 |
|
XM_017021108.1 |
XP_016876597.1
|
5, 6 |
761 |
|
XM_017021109.1 |
XP_016876598.1
|
5, 6 |
761 |
|
XM_017021110.1 |
XP_016876599.1
|
5, 6 |
761 |
|
XM_017021111.1 |
XP_016876600.1
|
5, 6 |
642 |
|
XM_017021112.1 |
XP_016876601.1
|
5, 6 |
529 |
|
GenBank mRNA |
AB011174.1 |
- |
5, 6 |
767 |
|
AY320284.2 |
- |
5, 6 |
1095 |
|
BC010663.1 |
- |
2, 3 |
203 |
|
BC050351.1 |
- |
5, 6 |
634 |
5264871 |
BC065220.1 |
- |
5, 6 |
709 |
5732786 |
BC131591.1 |
- |
5, 6 |
620 |
|
View Full Product Details