Gene Transcripts

Gene Symbol
Mrc2
Entrez Gene ID
498011
Gene Name
mannose receptor, C type 2
Gene Aliases
Endo180
Chromosome Location
Chr. 10: 93520311 - 93581599 on Build Rnor_6.0
UniGene ID
Rn.213511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024687.1 NP_001019858.1 10 1616
XM_006247631.2 XP_006247693.1 10 1741
GenBank mRNA DQ058624.1 - 10 1616

View Full Product Details