Gene Transcripts
- Gene Symbol
- TRAPPC10
- Entrez Gene ID
- 7109
- Gene Name
- trafficking protein particle complex 10
- Gene Aliases
- EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
- Chromosome Location
- Chr. 21: 44012325 - 44106552 on Build GRCh38
- UniGene ID
- Hs.126221
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003274.4 |
NP_003265.3
|
3, 4 |
452 |
|
XM_005261168.4 |
XP_005261225.1
|
3, 4 |
445 |
|
XM_011529714.2 |
XP_011528016.1
|
3, 4 |
445 |
|
XM_011529715.2 |
XP_011528017.1
|
3, 4 |
445 |
|
XM_011529716.2 |
XP_011528018.1
|
3, 4 |
445 |
|
XM_011529717.2 |
XP_011528019.1
|
3, 4 |
445 |
|
XM_011529718.2 |
XP_011528020.1
|
3, 4 |
445 |
|
XM_011529719.2 |
XP_011528021.1
|
3, 4 |
445 |
|
XM_011529720.2 |
XP_011528022.1
|
3, 4 |
445 |
|
XM_011529725.2 |
XP_011528027.1
|
3, 4 |
445 |
|
XM_011529726.2 |
XP_011528028.1
|
3, 4 |
445 |
|
XM_011529730.2 |
XP_011528032.1
|
3, 4 |
445 |
|
XM_017028454.1 |
XP_016883943.1
|
3, 4 |
445 |
|
GenBank mRNA |
AL559301.3 |
- |
3, 4 |
459 |
|
BC046241.1 |
- |
3, 4 |
430 |
|
BC052247.1 |
- |
3, 4 |
430 |
|
BC094823.1 |
- |
3, 4 |
430 |
|
BC101728.1 |
- |
3, 4 |
372 |
8069234 |
BF979287.1 |
- |
3, 4 |
469 |
|
U19252.1 |
- |
3, 4 |
420 |
|
U61500.1 |
- |
3, 4 |
436 |
|
View Full Product Details