Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022410.3 | NP_071855.2 | 3 | 642 | |
GenBank mRNA | AJ312390.1 | - | 3 | 566 | |
AK040822.1 | - | 3 | 573 | ||
AK053221.1 | - | 3 | 578 | ||
AK083071.1 | - | 3 | 572 | ||
AK147203.1 | - | 3 | 565 | ||
AK147206.1 | - | 3 | 574 | ||
AK147208.1 | - | 3 | 582 | ||
AK147209.1 | - | 3 | 573 | ||
AK147210.1 | - | 3 | 573 | ||
AK147211.1 | - | 3 | 617 | ||
AK147215.1 | - | 3 | 560 | ||
AK147216.1 | - | 3 | 515 | ||
AK147221.1 | - | 3 | 520 | ||
AK147222.1 | - | 3 | 561 | ||
AK147223.1 | - | 3 | 566 | ||
AK147233.1 | - | 3 | 573 | ||
AK147235.1 | - | 3 | 573 | ||
AK147296.1 | - | 3 | 573 | ||
AK147407.1 | - | 3 | 576 | ||
AK147430.1 | - | 3 | 583 | ||
AK165379.1 | - | 3 | 523 | ||
BC116417.1 | - | 3 | 527 | ||
BC116418.1 | - | 3 | 527 | ||
BC128497.1 | - | 3 | 527 | ||
BC128498.1 | - | 3 | 527 |