Gene Transcripts

Gene Symbol
C5orf49
Entrez Gene ID
134121
Gene Name
chromosome 5 open reading frame 49
Gene Aliases
Chromosome Location
Chr. 5: 7830378 - 7851490 on Build GRCh38
UniGene ID
Hs.709979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001089584.2 NP_001083053.1 3 2136
GenBank mRNA BC062569.1 - 3 2136

View Full Product Details