Gene Transcripts

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
Chromosome Location
Chr. 12: 42081845 - 42144871 on Build GRCh38
UniGene ID
Hs.259347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099650.1 NP_001093120.1 2 473
NM_173601.1 NP_775872.1 3 566
XM_017019211.1 XP_016874700.1 3 305
GenBank mRNA AK125141.1 - 2 326
BC030023.1 - 3 574 5266607
BC039145.1 - 2 313 4508300

View Full Product Details