Gene Transcripts

Gene Symbol
NOTO
Entrez Gene ID
344022
Gene Name
notochord homeobox
Gene Aliases
Chromosome Location
Chr. 2: 73202258 - 73211212 on Build GRCh38
UniGene ID
Hs.694384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134462.1 NP_001127934.1 1 728
GenBank mRNA AK295936.1 - 1 728
AK316410.1 - 1 684

View Full Product Details