Gene Transcripts

Gene Symbol
PTPRQ
Entrez Gene ID
374462
Gene Name
protein tyrosine phosphatase, receptor type Q
Gene Aliases
DFNB84, DFNB84A, PTPGMC1, R-PTP-Q
Chromosome Location
Chr. 12: 80424261 - 80680273 on Build GRCh38
UniGene ID
Hs.539284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145026.1 NP_001138498.1 9 1292
XM_017019273.1 XP_016874762.1 13 2094
XM_017019274.1 XP_016874763.1 13 2095
XM_017019275.1 XP_016874764.1 13 2095
GenBank mRNA DV080649.1 - 2 243

View Full Product Details