Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002612.3 | NP_002603.1 | 4 | 737 | |
GenBank mRNA | AK096428.1 | - | 5 | 536 | |
AK223328.1 | - | 4 | 633 | ||
AK312424.1 | - | 4 | 643 | ||
BC040239.1 | - | 4 | 617 | 3047987 | |
U54617.1 | - | 4 | 639 |