Gene Transcripts
- Gene Symbol
- RP1
- Entrez Gene ID
- 6101
- Gene Name
- retinitis pigmentosa 1 (autosomal dominant)
- Gene Aliases
- DCDC4A, ORP1
- Chromosome Location
- Chr. 8: 54509448 - 54630834 on Build GRCh38
- UniGene ID
- Hs.732820
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK304443.1 |
- |
19 |
2278 |
|
AK316511.1 |
- |
19 |
2712 |
|
- Gene Symbol
- LOC107984125
- Entrez Gene ID
- 107984125
- Gene Name
- lipoxygenase homology domain-containing protein 1-like
- Gene Aliases
- Chromosome Location
- Chr. 8: 54657854 - 54871720 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details