Gene Transcripts

Gene Symbol
TBC1D19
Entrez Gene ID
55296
Gene Name
TBC1 domain family member 19
Gene Aliases
Chromosome Location
Chr. 4: 26583924 - 26858919 on Build GRCh38
UniGene ID
Hs.479403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292054.1 NP_001278983.1 4, 5 563
NM_018317.3 NP_060787.2 7, 8 758
XM_011513852.2 XP_011512154.1 7, 8 598
XM_017008363.1 XP_016863852.1 7, 8 598
XM_017008364.1 XP_016863853.1 7, 8 598
GenBank mRNA AB449905.1 - 4, 5 293
AK001944.1 - 7, 8 598
AK225395.1 - 7, 8 598
AK289631.1 - 7, 8 598
AK310795.1 - 7, 8 605
BC031642.1 - 7, 8 554 5175317

View Full Product Details