Gene Transcripts
- Gene Symbol
- WDR33
- Entrez Gene ID
- 55339
- Gene Name
- WD repeat domain 33
- Gene Aliases
- NET14, WDC146
- Chromosome Location
- Chr. 2: 127704234 - 127811187 on Build GRCh38
- UniGene ID
- Hs.554831
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006622.2 |
NP_001006623.1
|
3, 4 |
466 |
|
NM_001006623.2 |
NP_001006624.1
|
3, 4 |
466 |
|
NM_018383.4 |
NP_060853.3
|
3, 4 |
466 |
|
XM_005263697.2 |
XP_005263754.1
|
3, 4 |
466 |
|
XM_011511436.1 |
XP_011509738.1
|
3, 4 |
495 |
|
XM_017004435.1 |
XP_016859924.1
|
3, 4 |
466 |
|
XM_017004436.1 |
XP_016859925.1
|
3, 4 |
466 |
|
GenBank mRNA |
AB044749.1 |
- |
3, 4 |
426 |
|
AK002156.1 |
- |
3, 4 |
441 |
|
AK223319.1 |
- |
3, 4 |
443 |
|
BC005401.1 |
- |
3, 4 |
450 |
|
BC013990.2 |
- |
3, 4 |
441 |
|
BC068484.1 |
- |
3, 4 |
441 |
|
BU597855.1 |
- |
3, 4 |
453 |
|
DA768238.1 |
- |
3, 4 |
466 |
|
View Full Product Details