Gene Transcripts
- Gene Symbol
- OVOS
- Entrez Gene ID
- 408186
- Gene Name
- ovostatin
- Gene Aliases
- Chromosome Location
- Chr. 12: 9517803 - 9573287 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647938.1 |
- |
5 |
1298 |
|
View Full Product Details