Gene Transcripts

Gene Symbol
OVOS
Entrez Gene ID
408186
Gene Name
ovostatin
Gene Aliases
Chromosome Location
Chr. 12: 9517803 - 9573287 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX647938.1 - 12 6756

Gene Symbol
OVOS2
Entrez Gene ID
144203
Gene Name
ovostatin 2
Gene Aliases
Chromosome Location
Chr. 12: 31114623 - 31206154 on Build GRCh38
UniGene ID
Hs.524331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CR749395.1 - 5 2727

View Full Product Details