Gene Transcripts

Gene Symbol
PPM1H
Entrez Gene ID
57460
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1H
Gene Aliases
ARHCL1, NERPP-2C, URCC2
Chromosome Location
Chr. 12: 62643982 - 62935135 on Build GRCh38
UniGene ID
Hs.435479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020700.1 NP_065751.1 10 3914
XM_011538578.2 XP_011536880.1 10 4050
XM_011538579.1 XP_011536881.1 10 3602
GenBank mRNA AB032983.1 - 9 3508
AB084258.1 - 10 4179
BC157843.1 - 10 3926 9057062

View Full Product Details