Gene Transcripts

Gene Symbol
SRL
Entrez Gene ID
6345
Gene Name
sarcalumenin
Gene Aliases
Chromosome Location
Chr. 16: 4189374 - 4242253 on Build GRCh38
UniGene ID
Hs.10041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098814.1 NP_001092284.1 4 300
NM_001323667.1 NP_001310596.1 5 430
NM_001323668.1 NP_001310597.1 5 1678
XM_017023527.1 XP_016879016.1 5 1625
XM_017023528.1 XP_016879017.1 4 331
XM_017023529.1 XP_016879018.1 4 346
GenBank mRNA AK056588.1 - 4 300
AK302601.1 - 4 336
AL831973.2 - 4 272
AL832361.1 - 5 1672
BX647476.1 - 5 1672
DB149163.1 - 4 300

View Full Product Details