Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173516.1 | NP_001166987.1 | 5, 6 | 644 | |
NM_024597.3 | NP_078873.2 | 5, 6 | 756 | ||
XM_005262472.1 | XP_005262529.1 | 5, 6 | 611 | ||
XM_017029843.1 | XP_016885332.1 | 5, 6 | 608 | ||
GenBank mRNA | AK301478.1 | - | 5, 6 | 645 | |
BC040518.1 | - | 5, 6 | 620 | ||
BC064350.1 | - | 5, 6 | 559 |