Gene Transcripts

Gene Symbol
SHANK3
Entrez Gene ID
85358
Gene Name
SH3 and multiple ankyrin repeat domains 3
Gene Aliases
DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2
Chromosome Location
Chr. 22: 50674642 - 50733212 on Build GRCh38
UniGene ID
Hs.149035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033517.1 NP_277052.1 9 987
GenBank mRNA AB569469.1 - 9 987

View Full Product Details