Gene Transcripts

Gene Symbol
PEG10
Entrez Gene ID
23089
Gene Name
paternally expressed 10
Gene Aliases
EDR, HB-1, MEF3L, Mar2, Mart2, RGAG3
Chromosome Location
Chr. 7: 94656325 - 94669695 on Build GRCh38
UniGene ID
Hs.147492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040152.1 NP_001035242.1 2 2281
NM_001172437.2 NP_001165908.1 2 2292
NM_001172438.2 NP_001165909.1 2 2292
NM_001184961.1 NP_001171890.1 2 2281
NM_001184962.1 NP_001171891.1 2 2281
NM_015068.3 NP_055883.2 2 2281
GenBank mRNA AB028974.1 - 1 1854
AB049150.1 - 2 2060
AB049834.1 - 1 1919
AF216076.1 - 1 1812
AK299837.1 - 2247

View Full Product Details