Gene Transcripts

Gene Symbol
LRRC59
Entrez Gene ID
55379
Gene Name
leucine rich repeat containing 59
Gene Aliases
PRO1855, p34
Chromosome Location
Chr. 17: 50381233 - 50397553 on Build GRCh38
UniGene ID
Hs.370927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018509.3 NP_060979.2 7 2600
GenBank mRNA AF119857.1 - 4 1958
AK025328.1 - 7 2600
BC052279.1 - 7 2518 4127952

View Full Product Details