Gene Transcripts
- Gene Symbol
- PATL2
- Entrez Gene ID
- 197135
- Gene Name
- PAT1 homolog 2
- Gene Aliases
- Pat1a, hPat1a
- Chromosome Location
- Chr. 15: 44665732 - 44711389 on Build GRCh38
- UniGene ID
- Hs.11594
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145112.1 |
NP_001138584.1
|
7, 8 |
763 |
|
XM_011521336.2 |
XP_011519638.2
|
7, 8 |
1589 |
|
XM_011521337.2 |
XP_011519639.2
|
7, 8 |
1724 |
|
XM_011521338.2 |
XP_011519640.1
|
9, 10 |
1288 |
|
XM_011521339.2 |
XP_011519641.1
|
8, 9 |
1169 |
|
XM_011521340.2 |
XP_011519642.1
|
7, 8 |
1110 |
|
XM_011521341.1 |
XP_011519643.1
|
7, 8 |
873 |
|
XM_011521342.2 |
XP_011519644.1
|
5, 6 |
1770 |
|
XM_011521343.2 |
XP_011519645.1
|
8, 9 |
1120 |
|
XM_011521344.2 |
XP_011519646.1
|
5, 6 |
562 |
|
XM_011521345.2 |
XP_011519647.1
|
8, 9 |
1082 |
|
XM_011521346.2 |
XP_011519648.2
|
3, 4 |
1296 |
|
XM_011521347.1 |
XP_011519649.1
|
6, 7 |
678 |
|
XM_011521348.2 |
XP_011519650.1
|
6, 7 |
678 |
|
XM_017022000.1 |
XP_016877489.1
|
7, 8 |
1581 |
|
XM_017022001.1 |
XP_016877490.1
|
5, 6 |
710 |
|
GenBank mRNA |
BI835136.1 |
- |
6, 7 |
678 |
|
View Full Product Details