Gene Transcripts

Gene Symbol
RTL1
Entrez Gene ID
388015
Gene Name
retrotransposon-like 1
Gene Aliases
MART1, Mar1, PEG11
Chromosome Location
Chr. 14: 100880655 - 100884847 on Build GRCh38
UniGene ID
Hs.298602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134888.2 NP_001128360.1 1 2230
GenBank mRNA BC150617.1 - 1 2230

View Full Product Details