Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
54221
Gene Name
syntrophin gamma 2
Gene Aliases
G2SYN, SYN5
Chromosome Location
Chr. 2: 950868 - 1367991 on Build GRCh38
UniGene ID
Hs.595069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018968.3 NP_061841.2 15 1449
XM_011510366.2 XP_011508668.1 13 1486
XM_017004361.1 XP_016859850.1 15 1411
XM_017004362.1 XP_016859851.1 15 1417
XM_017004363.1 XP_016859852.1 15 1598
XM_017004364.1 XP_016859853.1 12 4739
XM_017004367.1 XP_016859856.1 16 1487
XM_017004373.1 XP_016859862.1 10 2053
GenBank mRNA AJ003029.1 - 15 1451
BC125251.1 - 9 997 40025232
BG208326.1 - 1 35

View Full Product Details