Gene Transcripts

Gene Symbol
SH2D5
Entrez Gene ID
400745
Gene Name
SH2 domain containing 5
Gene Aliases
Chromosome Location
Chr. 1: 20719731 - 20732640 on Build GRCh38
UniGene ID
Hs.591522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103160.1 NP_001096630.1 9 3150
NM_001103161.1 NP_001096631.1 10 3279
XM_011541459.1 XP_011539761.1 10 3744
XM_011541460.1 XP_011539762.1 11 3584
XM_011541461.2 XP_011539763.1 10 2857
XM_011541462.1 XP_011539764.1 10 3741
GenBank mRNA AK123236.1 - 1 1695
AK123841.1 - 6 2508
AK124869.1 - 9 3150

View Full Product Details