Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080429.2 | NP_001073898.1 | 18 | 3540 | |
NM_020902.1 | NP_065953.1 | 16 | 3459 | ||
XM_005272491.3 | XP_005272548.1 | 17 | 3476 | ||
XM_011528154.2 | XP_011526456.1 | 18 | 3459 | ||
XM_017027036.1 | XP_016882525.1 | 18 | 3513 | ||
GenBank mRNA | AB040976.1 | - | 9 | 2362 | |
AF289580.1 | - | 6 | 1412 | ||
AL833927.1 | - | 6 | 2142 | ||
BC020431.1 | - | 18 | 3383 | ||
BC128201.1 | - | 2 | 58 | ||
BC128202.1 | - | 2 | 58 |