Gene Transcripts

Gene Symbol
SRSF7
Entrez Gene ID
6432
Gene Name
serine and arginine rich splicing factor 7
Gene Aliases
9G8, AAG3, SFRS7
Chromosome Location
Chr. 2: 38743599 - 38751494 on Build GRCh38
UniGene ID
Hs.309090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031684.2 NP_001026854.1 3 582
NM_001195446.1 NP_001182375.1 3 582
XM_005264484.2 XP_005264541.1 3 449
XM_005264485.2 XP_005264542.1 3 449
GenBank mRNA AB445102.1 - 3 344
AK091425.1 - 3 397
AK225141.1 - 3 449
AK293667.1 - 3 449
AK297161.1 - 3 449
AY166860.1 - 3 470
AY513287.1 - 3 425
BC000997.2 - 3 429 2901284
BC017369.2 - 3 440 2967417
BC017908.1 - 3 447 4276642
BC022328.1 - 3 447 4251314
BT006745.1 - 3 344
L22253.1 - 3 449

View Full Product Details