Gene Transcripts
- Gene Symbol
- STAG3L2
- Entrez Gene ID
- 442582
- Gene Name
- stromal antigen 3-like 2 (pseudogene)
- Gene Aliases
- STAG3L1, STAG3L2P, STAG3L3
- Chromosome Location
- Chr. 7: 74881990 - 74890612 on Build GRCh38
- UniGene ID
- Hs.632310
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC002581.2 |
- |
10 |
1223 |
|
View Full Product Details