Gene Transcripts

Gene Symbol
Ly49si1
Entrez Gene ID
494206
Gene Name
immunoreceptor Ly49si1
Gene Aliases
Chromosome Location
Chr. 4: 163736176 - 163851595 on Build Rnor_6.0
UniGene ID
Rn.129033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009497.1 NP_001009497.1 3 248
XM_008763312.1 XP_008761534.1 3 499
XM_008763313.1 XP_008761535.1 4 529
XM_008763314.1 XP_008761536.1 4 323
XM_008763315.1 XP_008761537.1 3 499
XM_008763316.1 XP_008761538.1 3 499
XM_008763317.1 XP_008761539.1 2 409
XM_008763318.1 XP_008761540.1 2 409
XM_008763319.1 XP_008761541.1 3 499
GenBank mRNA AY659935.1 - 3 248

View Full Product Details