Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019617.3 | NP_062563.3 | 5, 6 | 578 | |
GenBank mRNA | AB039886.1 | - | 5, 6 | 578 | |
AJ966789.1 | - | 5, 6 | 578 | ||
AL833013.1 | - | 1 | 2180 | ||
AY139184.1 | - | 5, 6 | 516 | ||
AY359050.1 | - | 5, 6 | 502 | ||
AY456961.1 | - | 5, 6 | 515 | ||
BC059778.1 | - | 5, 6 | 504 |