Gene Transcripts

Gene Symbol
Prkx
Entrez Gene ID
501563
Gene Name
protein kinase, X-linked
Gene Aliases
Chromosome Location
Chr. X: 45080963 - 45124270 on Build Rnor_6.0
UniGene ID
Rn.16741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033963.1 NP_001029135.1 9 3239
GenBank mRNA BC091203.1 - 9 3239

View Full Product Details