Gene Transcripts

Gene Symbol
Popdc3
Entrez Gene ID
641520
Gene Name
popeye domain containing 3
Gene Aliases
Chromosome Location
Chr. 20: 50394592 - 50422551 on Build Rnor_6.0
UniGene ID
Rn.178910
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037369.1 NP_001032446.1 5 1119
XM_006256599.2 XP_006256661.1 4 1359
GenBank mRNA BC105761.1 - 5 1119
FQ218039.1 - 5 1022

View Full Product Details