Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252413.1 | NP_001239342.1 | 6 | 844 | |
NM_001252414.1 | NP_001239343.1 | 4 | 668 | ||
NM_001252415.1 | NP_001239344.1 | 7 | 891 | ||
NM_027186.2 | NP_081462.1 | 7 | 985 | ||
GenBank mRNA | AK010289.1 | - | 7 | 714 | |
BC047996.1 | - | 7 | 972 | 6509910 | |
BY576068.1 | - | 2 | 195 |