Gene Transcripts

Gene Symbol
Prss32
Entrez Gene ID
69814
Gene Name
protease, serine 32
Gene Aliases
2010001P08Rik, mT5, tryptase-5
Chromosome Location
Chr. 17: 23853772 - 23859776 on Build GRCm38
UniGene ID
Mm.244143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027220.2 NP_081496.2 5 1140
GenBank mRNA AY266139.1 - 5 1118
BC024903.1 - 5 1131 4208033

View Full Product Details