Gene Transcripts

Gene Symbol
Tex22
Entrez Gene ID
75671
Gene Name
testis expressed gene 22
Gene Aliases
1700028O09Rik, AV210422, Tep22
Chromosome Location
Chr. 12: 113074469 - 113088917 on Build GRCm38
UniGene ID
Mm.23377
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029381.1 NP_083657.1 3 1209
GenBank mRNA AF522069.1 - 3 1209
AK006471.1 - 3 677
BC048634.1 - 3 642 6770610

View Full Product Details