Gene Transcripts

Gene Symbol
Enpp7
Entrez Gene ID
238011
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Gene Aliases
Alk-SMase, Gm254
Chromosome Location
Chr. 11: 118988108 - 118993270 on Build GRCm38
UniGene ID
Mm.244114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030291.1 NP_001025462.1 3 741
GenBank mRNA AK144474.1 - 3 744
AK167678.1 - 3 744
BC141245.1 - 3 826 9055857
BC171967.1 - 3 826 9054253

View Full Product Details