Gene Transcripts

Gene Symbol
OR10H3
Entrez Gene ID
26532
Gene Name
olfactory receptor family 10 subfamily H member 3
Gene Aliases
Chromosome Location
Chr. 19: 15741393 - 15742343 on Build GRCh38
UniGene ID
Hs.553524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013938.1 NP_039226.1 1 583
GenBank mRNA BC106707.2 - 1 583
BC113844.1 - 1 583 40004027

View Full Product Details