Gene Transcripts

Gene Symbol
STAG3L2
Entrez Gene ID
442582
Gene Name
stromal antigen 3-like 2 (pseudogene)
Gene Aliases
STAG3L1, STAG3L2P, STAG3L3
Chromosome Location
Chr. 7: 74881990 - 74890612 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040584.2 8 1408
GenBank mRNA BM311130.1 - 1 30

Gene Symbol
STAG3L3
Entrez Gene ID
442578
Gene Name
stromal antigen 3-like 3 (pseudogene)
Gene Aliases
STAG3L1, STAG3L2, STAG3L3P
Chromosome Location
Chr. 7: 72997295 - 73005943 on Build GRCh38
UniGene ID
Hs.632310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040582.2 9 1683
GenBank mRNA BC030509.1 - 5 960

View Full Product Details