Gene Transcripts

Gene Symbol
RNASEH2C
Entrez Gene ID
84153
Gene Name
ribonuclease H2 subunit C
Gene Aliases
AGS3, AYP1
Chromosome Location
Chr. 11: 65717673 - 65720938 on Build GRCh38
UniGene ID
Hs.718438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032193.3 NP_115569.2 4 692
GenBank mRNA AF346606.1 - 4 692
AK024627.1 - 4 549
AK225131.1 - 4 549
AK297660.1 - 4 462
BC023588.2 - 4 572 4763204
BM921525.1 - 4 561
L08647.1 - 3 421

View Full Product Details