Gene Transcripts

Gene Symbol
PKDCC
Entrez Gene ID
91461
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
SGK493, Vlk
Chromosome Location
Chr. 2: 42048021 - 42058528 on Build GRCh38
UniGene ID
Hs.408542
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138370.2 NP_612379.2 7 1811
GenBank mRNA BC007901.2 - 5 766
BC062988.1 - 5 766
BC094697.1 - 7 1219 30529646

View Full Product Details