Gene Transcripts

Gene Symbol
Sertad3
Entrez Gene ID
499108
Gene Name
SERTA domain containing 3
Gene Aliases
Chromosome Location
Chr. 1: 84280945 - 84284681 on Build Rnor_6.0
UniGene ID
Rn.22759
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017513.1 NP_001017513.1 2 415
XM_006228582.1 XP_006228644.1 2 1389
GenBank mRNA BC091229.1 - 2 415

View Full Product Details