Gene Transcripts

Gene Symbol
PBXIP1
Entrez Gene ID
57326
Gene Name
PBX homeobox interacting protein 1
Gene Aliases
HPIP
Chromosome Location
Chr. 1: 154944080 - 154956163 on Build GRCh38
UniGene ID
Hs.505806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317734.1 NP_001304663.1 9 1226
NM_001317735.1 NP_001304664.1 7 895
NM_020524.3 NP_065385.2 10 1315
GenBank mRNA AF221521.1 - 10 1264
AK022497.1 - 9 1185
AK023219.1 - 10 1256
AK300987.1 - 6 604
AK303413.1 - 7 839
AK303863.1 - 7 740
AK315551.1 - 10 1296
BC016852.1 - 10 1243 3900461

View Full Product Details