Gene Transcripts

Gene Symbol
Oxct2a
Entrez Gene ID
64059
Gene Name
3-oxoacid CoA transferase 2A
Gene Aliases
Oxct, Oxct2, Scot, Scot-t1
Chromosome Location
Chr. 4: 123321875 - 123323679 on Build GRCm38
UniGene ID
Mm.270287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022033.4 NP_071316.1 1 1764
GenBank mRNA AB022180.2 - 1 1702
AK077075.1 - 1 1717

View Full Product Details