Gene Transcripts

Gene Symbol
Mnd1
Entrez Gene ID
76915
Gene Name
meiotic nuclear divisions 1 homolog (S. cerevisiae)
Gene Aliases
2610034E18Rik, AI591601, Gaj
Chromosome Location
Chr. 3: 84087933 - 84156861 on Build GRCm38
UniGene ID
Mm.274829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029797.3 NP_084073.2 2 171
GenBank mRNA AK011664.1 - 2 81
BC027741.1 - 2 98 5035682
BY083009.1 - 2 172
BY710901.1 - 2 81

Gene Symbol
Mnd1-ps
Entrez Gene ID
100042415
Gene Name
Mnd1 retrotransposed pseudogene
Gene Aliases
Gm3833
Chromosome Location
Chr. 14: 9884095 - 9886868 on Build GRCm38
UniGene ID
Mm.235938
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK156325.1 - 1 1881

View Full Product Details