Gene Transcripts
- Gene Symbol
- Ulbp1
- Entrez Gene ID
- 77777
- Gene Name
- UL16 binding protein 1
- Gene Aliases
- A430108B07Rik, MULT1
- Chromosome Location
- Chr. 10: 7444873 - 7473637 on Build GRCm38
- UniGene ID
- Mm.442442
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084712.1 |
- |
|
2944 |
|
AK085119.1 |
- |
|
3008 |
|
View Full Product Details