Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr. 2: 120013028 - 120179121 on Build GRCh38
UniGene ID
Hs.369232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184937.1 NP_001171866.1 16, 17 1530
NM_020909.3 NP_065960.2 16, 17 1530
XM_006712651.1 XP_006712714.1 16, 17 1510
XM_006712652.1 XP_006712715.1 16, 17 1825
XM_006712653.1 XP_006712716.1 16, 17 1530
XM_011511529.2 XP_011509831.1 16, 17 1510
XM_017004567.1 XP_016860056.1 16, 17 1366
GenBank mRNA AB046768.1 - 9, 10 779
AK023019.1 - 16, 17 1481
BC054508.1 - 16, 17 1465 5502663

View Full Product Details