Gene Transcripts

Gene Symbol
Fbxw19
Entrez Gene ID
235612
Gene Name
F-box and WD-40 domain protein 19
Gene Aliases
E330023O18
Chromosome Location
Chr. 9: 109478507 - 109495863 on Build GRCm38
UniGene ID
Mm.133921
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177703.3 NP_808371.1 5 446
GenBank mRNA AK087808.1 - 5 447

View Full Product Details