Gene Transcripts

Gene Symbol
TRIM13
Entrez Gene ID
10206
Gene Name
tripartite motif containing 13
Gene Aliases
CAR, DLEU5, LEU5, RFP2, RNF77
Chromosome Location
Chr. 13: 49997007 - 50018467 on Build GRCh38
UniGene ID
Hs.436922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007278.2 NP_001007279.1 4 1259
NM_005798.4 NP_005789.2 3 1060
NM_052811.3 NP_434698.1 3 1145
NM_213590.2 NP_998755.1 2 1595
GenBank mRNA AF220127.1 - 3 1000
AF220128.1 - 1 838
AF241850.1 - 3 1018
AJ224819.1 - 2 945
AK001806.1 - 1 286
AK314496.1 - 3 1104
AL360196.1 - 3 1000
AY191002.1 - 3 1093
AY455758.1 - 2 1570
AY764035.1 - 4 1229
BC003579.1 - 3 1106 3538675
BC063407.1 - 2 1537 5212073

View Full Product Details