Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr. 21: 31671000 - 31732118 on Build GRCh38
UniGene ID
Hs.17255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145444.1 NP_001138916.1 11 1833
NM_001145445.1 NP_001138917.1 12 1878
NM_020706.2 NP_065757.1 12 1878
XM_005261017.2 XP_005261074.1 11 1682
XM_006724035.2 XP_006724098.1 12 1847
XM_006724036.2 XP_006724099.1 12 1847
XM_017028415.1 XP_016883904.1 12 1836
XM_017028416.1 XP_016883905.1 12 1721
XM_017028417.1 XP_016883906.1 11 1682
GenBank mRNA AB032998.1 - 6 866
AF023142.1 - 12 1482
AK025748.1 - 7 897
AK308406.1 - 11 1684
AL117417.1 - 7 886
AL834304.1 - 12 1658
BC014921.1 - 12 1855 3907610
BC043353.1 - 12 1835 6019414
BC052286.1 - 12 1709 5442227
BC064990.1 - 12 1709 5432277

View Full Product Details