Gene Transcripts
- Gene Symbol
- NME9
- Entrez Gene ID
- 347736
- Gene Name
- NME/NM23 family member 9
- Gene Aliases
- NM23-H9, TXL-2, TXL2, TXNDC6
- Chromosome Location
- Chr. 3: 138261437 - 138331785 on Build GRCh38
- UniGene ID
- Hs.660992
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178130.3 |
NP_835231.1
|
12 |
1737 |
|
GenBank mRNA |
AK094830.1 |
- |
12 |
1737 |
|
View Full Product Details